Thermo Scientific™ Hypersil™ ODS-2 Classical C18 HPLC Columns
High-efficiency columns with proven results and reproducibility
  • Suitable for Octadecyl silane (C18) phase
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil ODS-2 C18 Columns are manufactured under strict ISO 9001:2000 guidelines and provide a dependable L1 alternative. These columns are tested to confirm reproducibility and ruggedness required in QA and QC laboratories. Provide separations over a wide range of applications including polar compounds, vitamins, and amino acids. Stable in pH range of 2 to 8.
309,000.00 - ₹342,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific 31603-054630 ODS-2 (C18) Column - Length 50mm, 4.6mm ID, 3µm particle size 0189014 Mfr # 31603-054630
COMPARE
4.6 50 3
₹309,000.00INR / Each
Thermo Scientific 31605-254630 ODS-2 HPLC Column, C18, 5 µm, 250 mm L x 4.6 mm ID 0189041 Mfr # 31605-254630
COMPARE
4.6 250 5
₹342,000.00INR / Each