Thermo Scientific Finntip Flex Pipette Tips
Reduce the amount of force needed to attach and eject tips for lower repetitive strain injury (RSI)
  • Flex material ensures tighter seal with pipette for superior accuracy and precision
  • Slip-free rack opens easily with one hand and simplifies repetitive pipetting and frequent tip attachment
12,500.00 - ₹52,200.00INR / Pkg of 960
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of
Availability Pricing
Thermo Scientific Finntip 94060510 Pipette Tips, 300 µL; 10 x 96 Racks WW-25717-02 Mfr # 94060510
COMPARE
300 10 racks; 96 tips per rack
₹28,900.00INR / Pkg of 960
Thermo Scientific Finntip 94060513 Sterile Pipette Tips, 300 µL, 10 x 96 Racks WW-25717-04 Mfr # 94060513
COMPARE
300 10 racks; 96 tips per rack (sterile)
₹33,800.00INR / Pkg of 960
Thermo Scientific Finntip 94060517 Pipette Tip Refill Kit, 300 µL; 20 x 96 Racks WW-25717-08 Mfr # 94060517
COMPARE
300 20 racks; 96 tips per rack
₹49,000.00INR / Pkg of 96
Thermo Scientific Finntip 94060710 Pipette Tips, 1000 µL; 10 x 96 Racks WW-25717-12 Mfr # 94060710
COMPARE
1000 10 racks; 96 tips per rack
₹28,100.00INR / Pkg of 960
Thermo Scientific Finntip 94060713 Sterile Pipette Tips, 1000 µL; 10 x 96 Racks WW-25717-14 Mfr # 94060713
COMPARE
1000 10 racks; 96 tips per rack (sterile)
₹33,800.00INR / Pkg of 960
Thermo Scientific Finntip 94060716 Pipette Tip Starter Kit, 1000 µL; 17 x 96 Racks WW-25717-16 Mfr # 94060716
COMPARE
1000 16 racks; 96 tips per rack with 1 reusable box
₹49,300.00INR / Pkg of 96
Thermo Scientific Finntip 94060717 Pipette Tip Refill Kit, 1000 µL; 16 x 96 Racks WW-25717-18 Mfr # 94060717
COMPARE
1000 16 racks; 96 tips per rack
₹46,900.00INR / Pkg of 96
Thermo Scientific Finntip 94060820 Pipette Tips, 1000 µL; 1000/Bg WW-25717-20 Mfr # 94060820
COMPARE
1000 Bag of 1000
₹21,600.00INR / Bag of 1000
Thermo Scientific Finntip 94060720 Pipette Tips, 1200 µL; 1000/Bg WW-25717-22 Mfr # 94060720
COMPARE
1200 Bag of 1000
₹21,100.00INR / Bag of 1000
Thermo Scientific Finntip 94060810 Pipette Tips, 1200 µL; 10 x 96 Racks WW-25717-24 Mfr # 94060810
COMPARE
1200 10 racks; 96 tips per rack
₹42,200.00INR / Pkg of 960
Thermo Scientific Finntip 94060813 Sterile Pipette Tips, 1200 µL; 10 x 96 Racks WW-25717-26 Mfr # 94060813
COMPARE
1200 10 racks; 96 tips per rack (sterile)
₹52,200.00INR / Pkg of 960
Thermo Scientific Finntip 94060970 Extended Pipette Tips, 10mL; 100/Bg WW-25717-28 Mfr # 94060970
COMPARE
10000 Bag of 100
₹12,500.00INR / Bag of 100
Showing 1- 12 of 12