Thermo Scientific Finnpipette F1 Single-channel Pipettors
A new level of sophistication created to meet the demands of modern laboratories
  • The first pipette to have an antimicrobial surface treatment to prevent contamination
  • Adjustable finger rest for a customized feel based on your preferences
  • Large, easy-to-read digital display on adjustable-volume models
  • Comfortable, lightweight design with smooth pipetting action
  • Soft-tip ejection
  • Color-coded plunger for quick and easy identification
  • 5-year warranty with web registration
75,800.00 - ₹105,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (µL) Min Volume (µL)
Availability Pricing
Thermo Scientific Finnpipette 4641030 Pipette , 1-10 uL, Pink; Each WW-25014-06 Mfr # 4641030N
COMPARE
Pink 10 1
₹105,000.00INR / Each
Thermo Scientific Finnpipette 4641100 Pipette , 100-1000 uL, Blue; Each WW-25014-20 Mfr # 4641100N
COMPARE
Blue 1000 100
₹105,000.00INR / Each
Thermo Scientific Finnpipette 4641110 Pipette , 0.5-5 mL, Green; Each WW-25014-22 Mfr # 4641110N
COMPARE
Green 5000 500
₹105,000.00INR / Each
Thermo Scientific Finnpipette 4641120 Pipette , 1-10 mL, Red; Each WW-25014-24 Mfr # 4641120N
COMPARE
Red 10000 1000
₹105,000.00INR / Each
Thermo Scientific Finnpipette 4651100 Pipette , Fixed at 3 mL; Each WW-25014-70 Mfr # 4651100N
COMPARE
3000
₹75,800.00INR / Each