Thermo Scientific Barnstead Smart2Pure Water Purification Systems
Both Type I and Type II water in a complete space-saving design
  • Display can be tilted for optimal reading
  • Variable-flow control for dispensing of Type I water
  • Quick-change cartridge design simplifies self maintenance, even in mid-operations
Barnstead Smart2Pure systems deliver up to 72 L/day of Type I ultrapure water or Type II pure water from a tap feed water supply. Depending on your requirements, choose from three production capacities—3, 6, or 12 L/hr. The 12 L/hr systems let you safely store high purity water in either a 30- or 60-L reservoir, easily accessible when you need it!
These compact systems are fully customizable to support your tissue culture, PCR, electrophoresis, atomic absorption, ICP, ICP/MS, and chromatography needs. In addition, applications requiring Type II water such as reagent preparation and glassware rinsing can be met. The RO membrane with pretreatment cartridge is separate from the DI cartridge, ensuring you only replace each cartridge as needed.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Production (L/hour) With Ultrafilter With UV Lamp
Availability Pricing
Thermo Scientific Barnstead Smart2Pure 3 UV Water Purification System, 3L/Hr WW-99259-02 Mfr # 50129872
COMPARE
3 No Yes
Call For Price
Thermo Scientific Barnstead Smart2Pure 3 UV/UF Water Purification System, 3L/Hr WW-99259-04 Mfr # 50129688
COMPARE
3 Yes Yes
Call For Price
Thermo Scientific Barnstead Smart2Pure 6 UV Water Purification System, 6L/Hr WW-99259-12 Mfr # 50129885
COMPARE
6 No Yes
Call For Price
Thermo Scientific Barnstead Smart2Pure 6 UV/UF Water Purification System, 6L/Hr WW-99259-14 Mfr # 50129887
COMPARE
6 Yes Yes
Call For Price
Thermo Scientific Barnstead Smart2Pure 12 UV Water Purification System, 12L/Hr WW-99259-22 Mfr # 50129890
COMPARE
12 No Yes