Thermo Scientific Barnstead Post Treatment Cartridges
Large surface area for long life
  • Easily install and remove cartridges with the quarter twist release canister
  • Flow rates of up to 2L/min.
Use post-treatment cartridges at the point of use for filtration of ultrapure water. Compatible with the Thermo Scientific™ Barnstead™ B-Pure™ Water Purification Systems, models 99253-10, 99253-15, 99253-00, and 99259-00. Purification systems sold separately.
123,000.00 - ₹127,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Pore size (µm)
Availability Pricing
Thermo Scientific Barnstead FL583X6 Posttreatment cartridges, Pore size: 0.2 µm 0150432 Mfr # FL583X6
COMPARE
Posttreatment cartridges, Pore size: 0.2 µm 0.2
₹127,000.00INR / Each
Thermo Scientific Barnstead FL583X5 Posttreatment cartridges, Pore size: 0.45 µm 0150434 Mfr # FL583X5
COMPARE
Posttreatment cartridges, Pore size: 0.45 µm 0.45
₹123,000.00INR / Each