Thermo Scientific Barnstead Bantam Deionizer
View resistivity reading directly from unit
  • Selectable cartridges to best meet your application
Use this nonpressure cartridge system for polishing pretreated water or single-stage deionization
of tap water. Deionizer monitor indicates when it is time to change cartridges. When the monitor
drops below the acceptable minimum purity, or when it registers in the 0.05 to 0.025 MΩ range, the cartridge should be replaced. Monitor shows resistivity of effluent from 25 to 18 MΩ-cm.
Disposable cartridges (sold separately) are easily replaced by turning a thumbscrew.
Deionizer mounts on benchtop or wall.
432,000.00 - ₹436,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific Barnstead D0800 Bantam Deionizer, 120V 9926101 Mfr # D0800
COMPARE
120
₹436,000.00INR / Each
Thermo Scientific D0805 Bantam Deionizer, 240V 9926103 Mfr # D0805
COMPARE
240
₹432,000.00INR / Each