Thermo Scientific AquaPro 1/2-DIN Process Analyzer
Flexibility platform changes from one to up to four sensors with parameter modules
  • User-friendly menu in your choice of seven languages
  • Backlit, color displaywith prompts in user-selectable languages
  • Two 2-to-20 mA current outputs per sensor
  • Optional digital communication protocols available
  • Call to configure multiple parameters for up to four channels
The AquaPro is an intelligent multi-input process analyzer designed for a wide range of in-line applications. Analyzer measures key parameters simultaneously on a large 4 ¼” color display. Color error-warning flags are visible in any lighting. Straightforward and intuitive menu operation offers plain language prompts in seven user-selectable languages--English, Spanish, French, German, Italian, Portuguese, and Chinese.
Analyzer comes equipped with pH, ORP, conductivity, toroidal conductivity, or dissolved oxygen measurements modules. For temperature-compensated measurements, use with 1000 Pt RTD sensors (not included; see pages 0000-0000). For added versatility, call our Application Specialists to purchase additional modules for multi-parameter measurements on up to four channels.
Durable UV-resistant case with NEMA 4X and IP66 waterproof ratings can be pipe, panel or wall mounted. Analyzer also features a built-in USB data port that makes it easy to download up to 1000 data points, or use it to upload new software in the workplace. Easy interface to PLC, SCADA, and other PC-based control systems. Convenient add-on communication cards (sold separately below) help you adhere to current industry data communication protocols.
426,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific with PH module AquaPro Multiparameter Analyzer/with pH Module WW-56710-02 Mfr # AP1XXXK
COMPARE
AquaPro Multiparameter Analyzer/with pH Module
₹426,000.00INR / Each
Thermo Scientific AP3XXXK AquaPro Multiparameter Analyzer/ with Conductivity Module WW-56710-06 Mfr # AP3XXXK
COMPARE
AquaPro Multiparameter Analyzer/ with Conductivity Module
₹426,000.00INR / Each
Thermo Scientific with DO module AquaPro Multiparameter Analyzer/ with Polarographic DO Module WW-56710-10 Mfr # AP5XXXK
COMPARE
AquaPro Multiparameter Analyzer/ with Polarographic DO Module
₹426,000.00INR / Each