Thermo Scientific™ Accucore™ C18 HPLC Columns
High-speed, high-resolution separations
  • Solid core with porous outer layer allow fast and efficient separations without excessive back pressure
  • Controlled automated production processes result in reproducible yet rugged columns
  • Compatible with most HPLC instruments
These HPLC columns feature Core Enhanced Technology™ solid-core particles, reducing band broadening to improve resolution and sensitivity compared to fully porous particles. Tight control of particle size distribution creates a uniform and stable packed bed able to withstand temperature and pH extremes. Separation efficiency is comparable to < 2 µm fully-porous particles, but these columns do not generate comparable back pressure, so they are compatible with most standard HPLC systems.
Accucore C18 columns use hydrophobic interactions to separate a broad range of analytes and provide optimum retention of nonpolar compounds..
323,000.00 - ₹416,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific Accucore HPLC Column, C-18, 2.6 um Particle Size, 10 cm x 2.1 mm 0190551 Mfr # 17126-102130
COMPARE
HPLC Column, C-18, 2.6 um Particle Size, 10 cm x 2.1 mm
₹323,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 2.6 um Particle Size, 10 cm x 4.6 mm 0190553 Mfr # 17126-104630
COMPARE
HPLC Column, C-18, 2.6 um Particle Size, 10 cm x 4.6 mm
₹336,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 2.6 um Particle Size, 15 cm x 2.1 mm 0190554 Mfr # 17126-152130
COMPARE
HPLC Column, C-18, 2.6 um Particle Size, 15 cm x 2.1 mm
₹378,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 2.6 um Particle Size, 15 cm x 3 mm 0190555 Mfr # 17126-153030
COMPARE
HPLC Column, C-18, 2.6 um Particle Size, 15 cm x 3 mm
₹386,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 2.6 um Particle Size, 15 cm x 4.6 mm 0190556 Mfr # 17126-154630
COMPARE
HPLC Column, C-18, 2.6 um Particle Size, 15 cm x 4.6 mm
₹386,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 4.0 um Particle Size, 10 cm x 2.1 mm 0190557 Mfr # 74104-102130
COMPARE
HPLC Column, C-18, 4.0 um Particle Size, 10 cm x 2.1 mm
₹357,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 4.0 um Particle Size, 10 cm x 3 mm 0190558 Mfr # 74104-103030
COMPARE
HPLC Column, C-18, 4.0 um Particle Size, 10 cm x 3 mm
₹357,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 4.0 um Particle Size, 10 cm x 4.6 mm 0190559 Mfr # 74104-104630
COMPARE
HPLC Column, C-18, 4.0 um Particle Size, 10 cm x 4.6 mm
₹357,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 4.0 um Particle Size, 15 cm x 3 mm 0190561 Mfr # 74104-153030
COMPARE
HPLC Column, C-18, 4.0 um Particle Size, 15 cm x 3 mm
₹377,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 4.0 um Particle Size, 15 cm x 4.6 mm 0190562 Mfr # 74104-154630
COMPARE
HPLC Column, C-18, 4.0 um Particle Size, 15 cm x 4.6 mm
₹377,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 4.0 um Particle Size, 25 cm x 3 mm 0190564 Mfr # 74104-253030
COMPARE
HPLC Column, C-18, 4.0 um Particle Size, 25 cm x 3 mm
₹416,000.00INR / Each
Thermo Scientific Accucore HPLC Column, C-18, 4.0 um Particle Size, 25 cm x 4.6 mm 0190565 Mfr # 74104-254630
COMPARE
HPLC Column, C-18, 4.0 um Particle Size, 25 cm x 4.6 mm
₹397,000.00INR / Each
Showing 1- 12 of 12