Texwipe ESD Cleanroom Swabs
Eliminates risk of adhesive contamination
  • Thermal bond construction
  • No blooming carbon particles, metals, or ionic antistatic contaminants
  • Lot coded for traceability and quality control
  • Certified to ISO 9001:2015
The ESD-Safe cleanroom swabs feature Stat-Rite® handle, a patented dissipative polymer for clean ESD protection without contributing particulate contaminants. The head material is thermally bonded to the handle to eliminate the risk of adhesive contamination.
Easily apply and remove lubricants, adhesives, and other solutions in a critical clean environment with EDS concerns. The swabs are also chemical compatibility with a variety of solutions and solvents.
Cleanroom processed, the swabs are packaged in an anti-static bag. Packaging includes 500 swabs/bag, 5 inner bags of 100 swabs, 5 bags/case for a total 2,500 swabs/case. Choose from CleanFoam®, Alpha® polyester kit, or dual micro-tip pick.
10,000.00 - ₹32,600.00INR / Bag of 500
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Handle Length (in) Swab Total Length (InchShort) Tip Length (InchShort)
Availability Pricing
Texwipe TX750E ESD-Safe CleanFoam® Mini Compressed Swabs, Non-Sterile; 500/bag. 3367620 Mfr # TX750E
COMPARE
3.484 3 31/64 25/64
₹24,600.00INR / Bag of 500
Texwipe TX752E ESD-Safe CleanFoam® Medium Compressed Cleanroom Swabs, Non-Sterile; 500/bag. 3367621 Mfr # TX752E
COMPARE
3.169 3 9/16 25/64
₹32,600.00INR / Bag of 500
Texwipe TX753E ESD-Safe CleanFoam® Mini Swabs, Non-Sterile; 500/bag. 3367622 Mfr # TX753E
COMPARE
1.969 2 3/4 25/32
₹24,000.00INR / Bag of 500
Texwipe TX757E ESD-Safe CleanFoam® Micro Swabs, Non-Sterile; 500/Bag 3367623 Mfr # TX757E
COMPARE
2.224 2 19/32 3/8
₹23,400.00INR / Bag of 500
Texwipe TX758E ESD-Safe Alpha® Polyester Knit Micro Swabs, Non-Sterile; 500/bag. 3367624 Mfr # TX758E
COMPARE
3.177 3 9/16 25/64
₹26,500.00INR / Bag of 500
Texwipe TX761D ESD-Safe Alpha® Polyester Knit Swabs with Long Handle, Non-Sterile; 100/bag. 3367625 Mfr # TX761D
COMPARE
3.15 3 35/64 25/64
₹10,000.00INR / Bag of 100
Texwipe TX769E ESD-Safe Dual Micro-Tip Pick Swabs, Non-Sterile; 500/bag. 3367626 Mfr # TX769E
COMPARE
5.787 6 29/64 43/64
₹19,500.00INR / Bag of 500
Showing 1- 7 of 7