Testo CompactPro Thermohygrometers with Wireless Capability
Professional and reliable long-term use thermohygrometer
  • Meter requires the probes to be ordered separately from a impressive range of options
These meters have an impressive range of optional probes (meter does not come with a probe) that can measures humidity, temperature, moisture, dew point, and absolute pressure within seconds. Five probe inputs ensure maximum flexibility: two cable inputs and three wireless inputs. All inputs are displayed on a large, backlit display for easy viewing. A rugged designed rubberized housing offers protection against dirt and impact, also equipped with a convenient carrying strap and built-in mounting magnets. Function keys provide access to advanced features including differential, mean, min/max, hold, programmable data files, and user-defined profiles. Optional wireless printer 37486-12 provides documentation of all channels; meter can be set up for timed interval printing for long-term monitoring. Model 37803-37 includes all these features plus adds a 10,000 datalogging, USB capacity and ComSoft software for easy programming and data transferring. In addition to standard probes with a wire, this meter can be easily upgraded anytime to wireless probes for use up to a distance of 20 meters. A maximum of three wireless probes can be read and displayed by the meter. To add this feature use wireless upgrade kit 37486-02 (includes one wireless handle and module), along with a wireless probe head.
The high-pressure dew point (HPD) probe 37803-42 incorporates both a quick-connect fitting and integral sample chamber to make dew point for simple measuring in compressed air systems, dryers, medial and instrument air, and other applications where an accurate dew point measurement is needed.
Whats Included: Calibration documentation, and one 9 V battery.
130,000.00 - ₹184,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
Testo 0560 6351 CompactPro Thermohygrometer with Wireless Capability WW-37803-36 Mfr # 0560 6351
COMPARE
1 9V (included) 100 122
₹130,000.00INR / Each
Testo 0563 6352 CompactPro Thermohygrometer with Log WW-37803-37 Mfr # 0563 6352
COMPARE
1 9V (included) 100 122
₹184,000.00INR / Each