Tegam Portable Thermocouple Calibrators
Laboratory grade calibration in a compact, portable package
  • Excellent source accuracy of ±0.003% ±5 µV
  • Simultaneously source and measure
  • Ergonomic design allows for one-handed operation
  • Sealed keypad keeps the dirt out
  • 500-hour battery life
Easily perform accurate on-site temperature calibration and verification of thermocouples, temperature transmitters, process controllers, and displays with these temperature calibrators. These calibrators offer the accuracy of lab grade calibrators with the added convenience of portability. The calibrators compensate for the ambient temperature, allowing you to perform calibrations in various conditions.
The calibrators meet the demanding temperature calibration requirements yet are simple to use. Perform a 10/90% calibration with the push of a button; the “0/+25%” key allows for fast 5-point checks at 0, 25, 50, 75, and 100%. Use the probe offset feature to zero out temperature probe errors, significantly improving overall measurement uncertainty. Increase productivity, efficiency, and quality using real-time statistical data. Calibrators display min, max, average, range, and standard deviation on demand, providing critical information and helping to analyze manufacturing process to make immediate adjustment for maximum efficiency and quality.
These durable meters are built to withstand challenging environments so you can easily take it with you into the field or plant. Calibrators meet MIL-PRF-28800F for shock, drop, and vibration tests. The sealed keypad keeps dust and fluids out without compromising tactile feedback. Easily view the readings on a large backlit LCD. Three AA batteries provide up to 500 hours of battery life; auto-power off turns unit off after 20 minutes of nonuse.
272,000.00 - ₹553,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Logging Types Of Probes Accepted Description
Availability Pricing
Tegam 940A Temperature Calibrator, 4 Thermocouple Types and mV 7890936 Mfr # 940A
COMPARE
No Type J, Type K, Type T, Type E Temperature Calibrator, 4 Thermocouple Types and mV
₹272,000.00INR / Each
Tegam 945A Temperature Calibrator, 14 Thermocouple Types and mV 7890937 Mfr # 945A
COMPARE
No Type J, Type K, Type T, Type E, Type N, Type B, Type R, Type S, Type G, Type C, Type D, Type L, Type P, Type U Temperature Calibrator, 14 Thermocouple Types and mV
₹468,000.00INR / Each
Tegam 948A Wireless Datalogging Calibrator, 14 Thermocouple Types and mV 7890938 Mfr # 948A
COMPARE
Yes Type J, Type K, Type T, Type E, Type N, Type B, Type R, Type S, Type G, Type C, Type D, Type L, Type P, Type U Wireless Datalogging Temperature Calibrator, 14 Thermocouple Types and mV
₹553,000.00INR / Each