Taylor Mechanical Portion Control Food Scale
Reliable scale in a small and inexpensive package
  • NSF Approved
  • Removable platform
  • Shatterproof lens
The Taylor mechanical portion control scales features a rotating dial, which allows you to adjust for the tare weight of products or containers. Thanks to its removable stainless steel platform, the scale is easy to clean and sanitize between uses. The scale also features a stainless steel spring and corrosion resistant internal components that provide exception durability, impeccable accuracy, and unmatched longevity compared to other mechanical portion control scales.
14,200.00 - ₹39,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Capacity (lbs) Readability (g)
Availability Pricing
Taylor TS32 Mechanical SS Food Scale, 32 oz and 900gm 1101004 Mfr # TS32
COMPARE
900 2 5
₹16,400.00INR / Each
Taylor TS5 Mechanical SS Food Scale, 5Lb and 2.2kg 1101005 Mfr # TS5
COMPARE
2200 5 20
₹16,400.00INR / Each
Taylor TS25KL Mechanical SS Food Scale, 25Lb and 11kg 1101006 Mfr # TS25KL
COMPARE
11000 25 50
₹14,200.00INR / Each
Taylor THD50 Heavy-Duty Mechanical SS Food Scale, 50lb and 22kg 1101007 Mfr # THD50
COMPARE
22000 50 100
₹34,500.00INR / Each
Taylor THD32D Heavy-Duty Mechanical SS Food Scale, 32 oz and 900gm 1101008 Mfr # THD32D
COMPARE
900 2 2
₹39,800.00INR / Each
Taylor TS50 Heavy-Duty Mechanical SS Food Scale, 50 lb x 4 oz. Rotating Dial 1101009 Mfr # TS50
COMPARE
22679.6 50
₹16,100.00INR / Each