Taylor Analog Bimetal Food Service Meat Thermometers
No more under or overcooked meals
740.00 - ₹4,800.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
Taylor 5939N Analog Bimetal Meat Thermometer, NSF Listed WW-90000-51 Mfr # 5939
COMPARE
200 120 Analog Bimetal Meat Thermometer, NSF Listed
₹2,000.00INR / Each
Taylor 3504 TruTemp Series Analog Bimetal Meat Thermometer with FDA Temperture Guidelines on Dial WW-90001-09 Mfr # 3504
COMPARE
190 130 Analog Bimetal Meat Thermometer with FDA Temperture Guidelines on Dial
₹740.00INR / Each
Taylor 5937N Classic Series  Roast / Yeast Glass Thermometer with Stainless Steel Sabre WW-90001-10 Mfr # 5937N
COMPARE
185 105 Roast / Yeast Glass Thermometer with Stainless Steel Sabre
₹1,200.00INR / Each
Taylor 61054J Commercial Series Instant-Read Analog Bimetal Meat Thermometer WW-90001-14 Mfr # 61054J
COMPARE
220 0 Instant-Read Analog Bimetal Meat Thermometer
₹4,800.00INR / Each