Taylor 3504 TruTemp Series Analog Bimetal Meat Thermometer with FDA Temperture Guidelines on Dial
Representative image only.
Taylor 3504 TruTemp Series Analog Bimetal Meat Thermometer with FDA Temperture Guidelines on Dial
Taylor | Mfr: 3504| Item: 9000109
No more under or overcooked meals
₹1,200.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹1,200.00INR / Each
Buy It Together
Current Item
Taylor 3504 TruTemp Series Analog Bimetal Meat Thermometer with FDA Temperture Guidelines on Dial
Taylor 3504 TruTemp Series Analog Bimetal Meat Thermometer with FDA Temperture Guidelines on Dial
9000109
₹1,200.00INR / Each
+ =
Total Price 19900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 2" easy-to-read dial
  • 4 1/2" long steel stem
  • Temperature range from 130 degrees Fahrenheit to 190 degrees Fahrenheit
  • Sliding indicator for fast temperature reference
  • 1 year warranty
More About this Item
TruTemp® Meat Thermometer dial features a scale for various meats and functions best as a leave-in thermometer. Quick and precise readings ensure proper cooking temperatures are reached. Hand wash only.
1 Year WarrantyNSF international certified
Specifications
  • Min Temperature (° F)130
  • Max Temperature (° F)190
  • Accuracy2%
  • Dial Size (in)2
  • Stem Length (in)4.5
  • Stem Material304 Stainless Steel
  • DescriptionAnalog Bimetal Meat Thermometer with FDA Temperture Guidelines on Dial