SUPERLOK SUT Union Tee Fittings, Stainless Steel
Achieve leak-free, high-performance connections with SUPERLOK Tube Fittings, designed for reliability in even the most demanding applications
  • Double and single ferrule compression fittings for secure connections
  • Easy installation with minimal steps for fast assembly
  • High-quality stainless steel construction
  • Wide range of fitting types for versatile industrial applications
  • Gauge-able fittings ensure proper tightening and safety
  • Union tees connect tubing in a "T" configuration, with all connections being the same size
SUPERLOK Tube Fittings are engineered to provide the most reliable connections in industries where safety and performance are critical. These products are easy to install, durable, and resistant to leaks. Made from top-grade stainless steel, SUPERLOK fittings are ideal for high-pressure systems, gas applications, and more. Ensure secure installation with precise gaugeability and superior construction.
5,400.00 - ₹10,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Compression Size / Tube Od (Port 3) (in)
Availability Pricing
Superlok USA Union Tee, 1/16", Stainless Steel; 1/Each 6780160 Mfr # SUT-1-S316
COMPARE
1/16 1/16 1/16
₹9,500.00INR / Each
Superlok USA Union Tee, 1/8", Stainless Steel; 1/Each 6780161 Mfr # SUT-2-S316
COMPARE
1/8 1/8 1/8
₹5,900.00INR / Each
Superlok USA Union Tee, 1/4", Stainless Steel; 1/Each 6780162 Mfr # SUT-4-S316**
COMPARE
1/4 1/4 1/4
₹5,400.00INR / Each
Superlok USA Union Tee, 3/8", Stainless Steel; 1/Each 6780163 Mfr # SUT-6-S316
COMPARE
3/8 3/8 3/8
₹6,800.00INR / Each
Superlok USA Union Tee, 1/2", Stainless Steel; 1/Each 6780164 Mfr # SUT-8-S316
COMPARE
1/2 1/2 1/2
₹10,400.00INR / Each