Superlok USA Union Tee, 1/2", Stainless Steel; 1/Each
Representative image only.
Superlok USA Union Tee, 1/2", Stainless Steel; 1/Each
Superlok USA | Mfr: SUT-8-S316| Item: 6780164
Achieve leak-free, high-performance connections with SUPERLOK Tube Fittings, designed for reliability in even the most demanding applications
₹10,400.00INR / Each
₹10,400.00INR / Each
Product Description
Key Features
  • Double and single ferrule compression fittings for secure connections
  • Easy installation with minimal steps for fast assembly
  • High-quality stainless steel construction
  • Wide range of fitting types for versatile industrial applications
  • Gauge-able fittings ensure proper tightening and safety
  • Union tees connect tubing in a "T" configuration, with all connections being the same size
More About this Item
SUPERLOK Tube Fittings are engineered to provide the most reliable connections in industries where safety and performance are critical. These products are easy to install, durable, and resistant to leaks. Made from top-grade stainless steel, SUPERLOK fittings are ideal for high-pressure systems, gas applications, and more. Ensure secure installation with precise gaugeability and superior construction.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialStainless Steel
  • Number Of Ports3
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)1/2
  • Compression Size / Tube Od (Port 1) (mm)12.7
  • Compression Size / Tube Od (Port 2) (in)1/2
  • Compression Size / Tube Od (Port 2) (mm)12.7
  • Compression Size / Tube Od (Port 3) (in)1/2
  • Compression Size / Tube Od (Port 3) (mm)12.7
  • Flow PatternTee
  • Max Temperature (° F)375
  • Max Temperature (° C)191
  • DescriptionUnion Tee, 1/2", Stainless Steel