SUPERLOK SU Union Fittings, Stainless Steel
Achieve leak-free, high-performance connections with SUPERLOK Tube Fittings, designed for reliability in even the most demanding applications
  • Double and single ferrule compression fittings for secure connections
  • Easy installation with minimal steps for fast assembly
  • High-quality stainless steel construction
  • Wide range of fitting types for versatile industrial applications
  • Gauge-able fittings ensure proper tightening and safety
  • Union fittings connect identical-sized tubing
SUPERLOK Tube Fittings are engineered to provide the most reliable connections in industries where safety and performance are critical. These products are easy to install, durable, and resistant to leaks. Made from top-grade stainless steel, SUPERLOK fittings are ideal for high-pressure systems, gas applications, and more. Ensure secure installation with precise gaugeability and superior construction.
2,800.00 - ₹5,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Flow Pattern
Availability Pricing
Superlok USA Union, 1/16", Stainless Steel; 1/Each 6780150 Mfr # SU-1-S316
COMPARE
1/16 1/16 Straight through
₹3,900.00INR / Each
Superlok USA Union, 1/8", Stainless Steel; 1/Each 6780151 Mfr # SU-2-S316
COMPARE
1/8 1/8 Straight through
₹2,900.00INR / Each
Superlok USA Union, 1/4", Stainless Steel; 1/Each 6780152 Mfr # SU-4-S316**
COMPARE
1/4 1/4 Straight through
₹2,800.00INR / Each
Superlok USA Union, 3/8", Stainless Steel; 1/Each 6780153 Mfr # SU-6-S316
COMPARE
3/8 3/8 Straight through
₹3,800.00INR / Each
Superlok USA Union, 1/2", Stainless Steel; 1/Each 6780154 Mfr # SU-8-S316
COMPARE
1/2 1/2 Straight through
₹5,400.00INR / Each