Superlok USA Union, 1/2", Stainless Steel; 1/Each
Representative image only.
Superlok USA Union, 1/2", Stainless Steel; 1/Each
Superlok USA | Mfr: SU-8-S316| Item: 6780154
Achieve leak-free, high-performance connections with SUPERLOK Tube Fittings, designed for reliability in even the most demanding applications
₹5,400.00INR / Each
₹5,400.00INR / Each
Buy It Together
Current Item
Superlok USA Union, 1/2", Stainless Steel; 1/Each
Superlok USA Union, 1/2", Stainless Steel; 1/Each
6780154
₹5,400.00INR / Each
+ =
Total Price 8500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Double and single ferrule compression fittings for secure connections
  • Easy installation with minimal steps for fast assembly
  • High-quality stainless steel construction
  • Wide range of fitting types for versatile industrial applications
  • Gauge-able fittings ensure proper tightening and safety
  • Union fittings connect identical-sized tubing
More About this Item
SUPERLOK Tube Fittings are engineered to provide the most reliable connections in industries where safety and performance are critical. These products are easy to install, durable, and resistant to leaks. Made from top-grade stainless steel, SUPERLOK fittings are ideal for high-pressure systems, gas applications, and more. Ensure secure installation with precise gaugeability and superior construction.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialStainless Steel
  • Number Of Ports2
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)1/2
  • Compression Size / Tube Od (Port 1) (mm)12.7
  • Compression Size / Tube Od (Port 2) (in)1/2
  • Compression Size / Tube Od (Port 2) (mm)12.7
  • Flow PatternStraight through
  • Max Temperature (° F)375
  • Max Temperature (° C)191
  • DescriptionUnion, 1/2", Stainless Steel