Stovall Belly Dancer Shaker
Gentle yet thorough mixing & washing action
  • Unique, undulating motion provides optimal movement for multiple staining and washing applications in gel and blotting techniques
  • Constructed with chemically resistant materials
Combination of vertical and horizontal orbital motion allows rapid exchange of solvents when staining and destaining gels and ensures lower background blotting. The lightweight and extremely quiet shaker has strong suction feet for load stability. Trouble free construction contains no belts or pulleys, keeping it simple and durable. High quality shunt wound DC motor and control allow for a long life and dependable performance. Variable speed control, from 0 to 125 rpm, with no cogging at slow speeds. Collar adjustment varies platform angle from flat to 12 °pitch. Orbital throw is 1 inch. The open-sided platform accommodates oversize containers.
What's included: a platform, removable nonskid mat, 6-ft 110/115VAC power cord (no cord included for 220/240VAC model).
634,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Ibi Scientific BDRAA220S Belly Dancer Shaker, 220/240VAC 5165005 Mfr # BDRAA220S
COMPARE
220 / 240
₹634,000.00INR / Each