Stirrer Motor with Electronic Speed Controller
Choose a motor that works with your electrical requirements
  • Smooth rotation and repeatability
This stirrer motor features a totally enclosed 1⁄40-hp motor and two output shafts. Use the geared shaft when you need higher torque (5.4 in-lb) and lower speeds (333 rpm max); and the direct-drive shaft when you need higher speeds (4000 rpm max) and lower torque (0.26 in-lb). Each shaft measures 5⁄16" dia.
The versatile and reliable electronic motor controller features feedback circuitry that maintains speed even with viscosity changes. Filtered circuitry ensures smooth rotation for repeatability in homogenizing and greatly reduces motor temperature and noise. The reversing switch lets the motor shaft rotate in either direction and reverses pestle direction during homogenization. The controller is connected to the motor with a cord so you can place it away from corrosive environments.
433,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Motor hP Power (VAC)
Availability Pricing
Glas-Col 099C K5424CE Tissue Homogenizing System Motor, remote controller; 240 VAC (CE) 0436925 Mfr # 099C K5424CE
COMPARE
0.025 240
₹433,000.00INR / Each