Static-Free Desiccators
Protect your equipment from electrostatic discharge
Safely store instruments that are sensitive to electrostatic discharges. Sturdy stainless steel and glass construction prevents the static buildup inherent to plastic desiccators. The inner glass surfaces of the desiccator are coated with a transparent, static dissipative polyester film.
Desiccators also come with a grounding plug to prevent static discharge when indoor humidity levels fall below 40%. Adjustable shelves (two included) prevent wasted space inside desiccator. Valve lets you adjust gas flow. Both models accept 1¼4" ID tubing. Model 24000-00 has 12 shelf levels; model 24000-10 has 28 shelf levels.
1,391,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Chamber Volume (cu ft)
Availability Pricing
Boekel Scientific 134900 Static-Free Desiccator, 6.4 cu ft, 2 adjustable shelves 2400010 Mfr # 134900
COMPARE
6.4
₹1,391,000.00INR / Each