Speirs Robertson AMS and AMB Series Balance Isolation Platforms
Removes up to 90% of vibrations to improve performance
  • Sound-deadened layer construction removes surface vibrations
  • Seamless pharma-grade (Ra finish 0.3) stainless steel worktops are chemical resistant
  • Meets stringent cleaning requirements
These anti vibration platform for balances offer an unique Sorbothane isolator design that removes up to 98% of unwanted vibrations. The low natural frequency of the isolators built into the platform
ensures vibrations common in most laboratories are effectively removed, increasing instrument performance.
282,000.00 - ₹1,384,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Height (in) Width (in)
Availability Pricing
Speirs Robertson AMS-30x45 AMS Balance Platform, Sorbothane Isolators; 30x45 cm 1150004 Mfr # AMS-30X45
COMPARE
25 2 1/8 11 3/4
₹282,000.00INR / Each
Speirs Robertson AMS-40x50 AMS Balance Platform, Sorbothane Isolators; 40x50 cm 1150005 Mfr # AMS-40X50
COMPARE
25 2 1/8 15 3/4
₹369,000.00INR / Each
Speirs Robertson AMS-60x60 AMS Balance Platform, Sorbothane Isolators; 60x60 cm 1150006 Mfr # AMS-60X60
COMPARE
25 2 1/8 23 5/8
₹455,000.00INR / Each
Speirs Robertson AMB-30x45 AMB Balance Platform, Passive Air Isolators; 30x45 cm 1150007 Mfr # AMB-30X45-SS
COMPARE
100 2 1/8 11 3/4
₹1,278,000.00INR / Each
Speirs Robertson AMB-40x50 AMB Balance Platform, Passive Air Isolators; 40x50 cm 1150008 Mfr # AMB-40X50-SS
COMPARE
100 2 1/8 15 3/4
₹1,105,000.00INR / Each
Speirs Robertson AMB-60x60 AMB Balance Platform, Passive Air Isolators; 60x60 cm 1150009 Mfr # AMB-60X60-SS
COMPARE
100 2 1/8 23 5/8
₹1,384,000.00INR / Each