Speirs Robertson AMP and AMF Series Microscope Isolation Platforms

Maximize microscope image quality by reducing up to 99% of vibrations
  • Isolates out vibration frequencies above 6 Hz—well below building vibration frequencies
  • Eliminates surface vibrations with sound deadened construction
  • Stainless steel worktop meets stringent cleanliness requirements
  • Choice of passive air or active air isolators in a variety of platform sizes
Ideal for laboratory microscopes, including sensitive instruments such as laser confocal and spinning disk microscopes. Maintenance-free platforms are simply installed on a laboratory bench and the microscope placed on top.
1,057,000.00 - ₹1,301,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Height (in) Width (in)
Availability Pricing
Speirs Robertson AMP-60x60 AMP Microscope Platform, Passive Air Isolators; 60x60 cm 1150011 Mfr # AMP-60X60-SS
COMPARE
23 5/8 2 1/2 23 5/8
₹1,057,000.00INR / Each
Speirs Robertson AMP-75x75 AMP Microscope Platform, Passive Air Isolators; 75x75 cm 1150012 Mfr # AMP-75X75-SS
COMPARE
29 1/2 2 1/2 29 1/2
₹1,148,000.00INR / Each
Speirs Robertson AMF-60x60 AMF Microscope Platform, Active Air Isolators; 60x60 cm 1150014 Mfr # AMF-60X60-SS
COMPARE
23 5/8 2 1/2 23 5/8
₹1,301,000.00INR / Each
Speirs Robertson AMF-90x75 AMF Microscope Platform, Active Air Isolators; 90x75 cm 1150016 Mfr # AMF-90X75-SS
COMPARE
29 1/2 2 1/2 35 3/8
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat