Spectra/Por 2 Regenerated Cellulose (RC) Dialysis Tubing
Get gentle separation and purification of solutes
  • Available in a variety of diameters and flat widths
  • MWCO from 12,000 to 14,000
The RC Dialysis tubing is packaged dry to provide you with easy handling and storage. Available in 15 meter rolls to provide better economy for applications requiring larger amounts of tubing.
Most of the dry tubing rolls contain glycerin as a humectant which is easily removed by soaking 30 min in water.
91,600.00 - ₹125,000.00INR / Box of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por 2 RC Dialysis Tubing, 12-14 kD MWCO, 10mm Flat Width, 6.4mm dia; 50' L 0290019 Mfr # 132676
COMPARE
6.4 10 14000
₹91,600.00INR / Box of 1
Spectra Por 2 RC Dialysis Tubing, 12-14 kD MWCO, 25mm Flat Width, 15.9mm dia; 50' L 0290020 Mfr # 132678
COMPARE
15.9 25 14000
₹107,000.00INR / Box of 1
Spectra Por 2 RC Dialysis Tubing, 12-14 kD MWCO, 45mm Flat Width, 28.6mm dia; 50' L 0290021 Mfr # 132680
COMPARE
28.6 45 14000
₹91,600.00INR / Box of 1
Spectra Por 2 RC Dialysis Tubing, 12-14 kD MWCO, 120mm Flat Width, 76.4mm dia; 50' L 0290023 Mfr # 132684
COMPARE
76.4 120 14000
₹125,000.00INR / Box of 1