Spectra Por 2 RC Dialysis Tubing, 12-14 kD MWCO, 10mm Flat Width, 6.4mm dia; 50' L

Representative image only.

Spectra Por 2 RC Dialysis Tubing, 12-14 kD MWCO, 10mm Flat Width, 6.4mm dia; 50' L

Spectra Por | Mfr: 132676| Item: 0290019

Get gentle separation and purification of solutes

₹91,600.00INR / Box of 1

₹91,600.00INR / Box of 1
Product Description

Key Features

  • Available in a variety of diameters and flat widths
  • MWCO from 12,000 to 14,000
  • 6 month warranty

More About this Item

The RC Dialysis tubing is packaged dry to provide you with easy handling and storage. Available in 15 meter rolls to provide better economy for applications requiring larger amounts of tubing.
Most of the dry tubing rolls contain glycerin as a humectant which is easily removed by soaking 30 min in water.

6 Month Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Flat Width (mm)10
  • Diameter (mm)6.4
  • Length Per Pack (ft)50
  • Diameter (in)0.25
  • Description2 Membranes; flat width, 10mm; diameter, 6.4mm; length, 50'
  • Warranty6 month
Reviews
Q&A