Small Sample Adapter for Rotational Viscometers
Use the small sample adapters when measuring low fluid volumes of 8 to 13 mL. Adapters include a water jacket for attachment to an external circulating bath (not included). Easily install or remove sample chamber without disturbing operation of viscometer or circulating bath.
230,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Cup Number Max Viscosity Range (cP) Power (VAC)
Availability Pricing
Toshniwal Small Sample Adapter L range WW-15944-49 Mfr # SMALL SAMPLE ADAPTER L
COMPARE
₹230,000.00INR / Each
Toshniwal Small Sample Adapter R range WW-15944-69 Mfr # SMALL SAMPLE ADAPTER R
COMPARE
₹230,000.00INR / Each
Toshniwal Small Sample Adapter H range WW-15944-70 Mfr # SMALL SAMPLE ADAPTER H
COMPARE
₹230,000.00INR / Each