Toshniwal Small Sample Adapter L range
Representative image only.
Toshniwal Small Sample Adapter L range
Toshniwal | Mfr: SMALL SAMPLE ADAPTER L| Item: 1594449
₹185,000.00INR / Each
₹185,000.00INR / Each
Product Description
More About this Item
Use the small sample adapters when measuring low fluid volumes of 8 to 13 mL. Adapters include a water jacket for attachment to an external circulating bath (not included). Easily install or remove sample chamber without disturbing operation of viscometer or circulating bath.
Specifications
  • DescriptionSmall Sample Adapter L range