Sieve Cleaning Brushes
Keeps sieves clean for better separation
  • Will not damage or hinder performance of sieve
These round sieve brushes are ideal for cleaning fine and coarse mesh sieves. The nylon brush is to clean 120 mesh and finer while brass bristle brushes are more suited for 100 mesh and coarser. Proper maintenance and cleaning of sieves prevents damage to sieve openings, keeping them clean and free of particles and ensures longest possible in-spec service life and accurate results.
Each brush measures 6” long and the diameter of the bristle head is 2”.
7,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
W.S. Tyler 8577 Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles WW-59986-96 Mfr # 8577
COMPARE
Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
₹7,200.00INR / Each
W.S. Tyler 8576 Round Sieve Brush for 100 Mesh Sieves and Coarser, Brass Bristles WW-59986-97 Mfr # 8576
COMPARE
Round Sieve Brush for 100 Mesh Sieves and Coarser, Brass Bristles
₹7,200.00INR / Each