W.S. Tyler 8577 Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
Representative image only.
W.S. Tyler 8577 Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
W.S. Tyler | Mfr: 8577| Item: 5998696
Keeps sieves clean for better separation
₹10,100.00INR / Each
₹10,100.00INR / Each
Buy It Together
Current Item
W.S. Tyler 8577 Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
W.S. Tyler 8577 Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
5998696
₹10,100.00INR / Each
+ + =
Total Price 51300.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Will not damage or hinder performance of sieve
  • 3 month warranty
More About this Item
These round sieve brushes are ideal for cleaning fine and coarse mesh sieves. The nylon brush is to clean 120 mesh and finer while brass bristle brushes are more suited for 100 mesh and coarser. Proper maintenance and cleaning of sieves prevents damage to sieve openings, keeping them clean and free of particles and ensures longest possible in-spec service life and accurate results.
Each brush measures 6” long and the diameter of the bristle head is 2”.
Use this brush with fine mesh sieves, 120 mesh and finer. Brush has nylon bristles and wood handle.
3 Month Warranty
Specifications
  • Length (InchShort)6
  • Diameter (InchShort)2
  • DescriptionRound Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
  • Warranty3 Months