Shimpo Jar Mills

Grind and mix rough material either wet or dry
  • Exact milling with adjustable rollers
  • Accommodate 1 to 10 L jars
These heavy-duty jar mills enable precise milling with varying jar sizes from 1 to 10 L. Compact design is ideal for small-scale production. Powder-coated steel construction provides long-lasting durability. Order porcelain jars with balls separately.
439,000.00 - ₹540,000.00INR / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Jar Capacity (L) Speed (rpm) Description
Availability Pricing
Shimpo PTA-01 Jar Mill, Single Jar Capacity; 115 VAC WW-25302-97 Mfr # PTA-01
COMPARE
10 247 Jar Mill, Single Jar Capacity; 115 VAC
₹439,000.00INR / Each
Shimpo PTA-02 Jar Mill, Double Jar Capacity; 115 VAC WW-25302-98 Mfr # PTA-02
COMPARE
20 247 Jar Mill, Double Jar Capacity; 115 VAC
₹540,000.00INR / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat