Shimpo DT-735 Portable, Light Weight Stroboscopes with Rechargeable Battery
Rechargeable batteries allows unlimited measuring power
  • Lightweight aluminum construction allows for convenient one-handed operation
  • Repeatable accuaracy of ±0.01% of reading
The portable stroboscopes are designed with a robust, extruded aluminum housing with a repeatable accuracy of ±0.01% of reading. Other features include a continuous duty cycle, microprocessor, external trigger, synchronous output, flash timer control, highly polished focused reflector, and manual flash controller knob.
240,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Flash Range (fpm) Description
Availability Pricing
Shimpo DT-735-KIT Portable, Light Weight Stroboscope Kit with Rechargeable Battery, Spare Flash Tube, and Protective Carrying Case 2530532 Mfr # DT-735-KIT
COMPARE
5 Digit LED 35000 Portable, Light Weight Stroboscope Kit with Rechargeable Battery, Spare Flash Tube, and Protective Carrying Case
₹240,000.00INR / Each