Shimpo DT-735-KIT Portable, Light Weight Stroboscope Kit with Rechargeable Battery, Spare Flash Tube, and Protective Carrying Case
Representative image only.
Shimpo DT-735-KIT Portable, Light Weight Stroboscope Kit with Rechargeable Battery, Spare Flash Tube, and Protective Carrying Case
Shimpo | Mfr: DT-735-KIT| Item: 2530532
Rechargeable batteries allows unlimited measuring power
₹240,000.00INR / Each
₹240,000.00INR / Each
Buy It Together
Current Item
Shimpo DT-735-KIT Portable, Light Weight Stroboscope Kit with Rechargeable Battery, Spare Flash Tube, and Protective Carrying Case
Shimpo DT-735-KIT Portable, Light Weight Stroboscope Kit with Rechargeable Battery, Spare Flash Tube, and Protective Carrying Case
2530532
₹240,000.00INR / Each
+
Shimpo Replacement Flashtube for DT-735 and DT-311D
2530533
₹13,700.00INR / Each
=
Total Price 253700.00 ADD 2 ITEMS TO CART
Product Description
Includes
Spare flash tube and carrying case.
Key Features
  • Lightweight aluminum construction allows for convenient one-handed operation
  • Repeatable accuaracy of ±0.01% of reading
  • 1 year warranty
More About this Item
The portable stroboscopes are designed with a robust, extruded aluminum housing with a repeatable accuracy of ±0.01% of reading. Other features include a continuous duty cycle, microprocessor, external trigger, synchronous output, flash timer control, highly polished focused reflector, and manual flash controller knob.
1 Year Warranty
Specifications
  • Min Flash Range (fpm)40
  • Max Flash Range (fpm)35000
  • Flash Accuracy (fpm)±0.01% of reading
  • Display Type5 Digit LED
  • Battery TypeInternal Recharageable Battery
  • DescriptionPortable, Light Weight Stroboscope Kit with Rechargeable Battery, Spare Flash Tube, and Protective Carrying Case