Shimpo DT-501X Panel-Mount Tachometers
Monitor speed, rate and time all in one unit
  • Scalable programming gives flexibility to set up and monitor any process
  • Large six-digit, red LED display is easy to read from across the room
  • Max/min measurements store automatically into memory
  • Compact body depth of 3.6” (92 mm) allows mounting in tight spaces
  • Waterproof front cover meets NEMA 4X/IP66 specifications for plant wash-down
The fully scalable 1/8-DIN panel-mount tachometer can operate in tachometer, flowmeter, process time span and elapsed time modes allowing it to monitor various process speeds, rates and time applications when combined with a compatible process speed sensor. It is suitable in automation applications such as machining operation timing, conveyor speed, motor output, volumetric flowrate, and elapsed time of oven curing.
The unit accepts multiple sensor inputs such as NPN open collector, contact and square, or sine wave signals. Optional outputs (analog, relay, NPN open collector) are available for additional automation control. An added teaching function auto-scales the display by inputting the sensor’s signal; saving on manual programming time.
Other features include an auto zero function which zeros the display after final signal initiates a preprogrammed time countdown and a test mode to check operational performance of the display and keys. Integral sensor power source eliminates need to purchase a separate power supply.
43,500.00 - ₹120,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Display Type Description
Availability Pricing
Shimpo DT-501XA Panel Meter Tachometer, 100-240 VAC Powered WW-54500-00 Mfr # DT-501XA
COMPARE
6-digit LED, red, 0.87\" (22 mm) H Panel Meter Tachometer, 100-240 VAC
₹43,500.00INR / Each
Shimpo DT-501XD Panel Meter Tachometer, 9-35 VDC Powered WW-54500-01 Mfr # DT-501XD
COMPARE
6-digit LED, red, 0.87\" (22 mm) H Panel Meter Tachometer, 9-35 VDC
₹50,000.00INR / Each
Shimpo DT-501XA-FVT Panel Meter Tachometer, 100-240 VAC Powered, Analog Output with Terminal Block Connection WW-54500-02 Mfr # DT-501XA-FVT
COMPARE
6-digit LED, red, 0.87\" (22 mm) H Panel Meter Tachometer, 100-240 VAC, Analog Output with Terminal Block Connection
₹90,300.00INR / Each
Shimpo DT-501XA-CPT Panel Meter Tachometer, 100-240 VAC Powered, Relay Output WW-54500-03 Mfr # DT-501XA-CPT
COMPARE
6-digit LED, red, 0.87\" (22 mm) H Panel Meter Tachometer, 100-240 VAC, Relay Output
₹81,600.00INR / Each
Shimpo DT-501XA-CPT-FVC Panel Meter Tachometer, 100-240 VAC Powered, Relay Output, Analog Output with 36 Pin Connection WW-54500-05 Mfr # DT-501XA-CPT-FVC
COMPARE
6-digit LED, red, 0.87\" (22 mm) H Panel Meter Tachometer, 100-240 VAC, Relay Output, Analog Output with 36 Pin Connection
₹120,000.00INR / Each
Shimpo DT-501XD-FVT Panel Meter Tachometer, 9-35 VDC Powered, Analog Output with Terminal Block Connection WW-54500-07 Mfr # DT-501XD-FVT
COMPARE
6-digit LED, red, 0.87\" (22 mm) H Panel Meter Tachometer, 9-35 VDC, Analog Output with Terminal Block Connection
₹96,800.00INR / Each
Shimpo DT-501XD-CPT Panel Meter Tachometer, 9-35 VDC Powered, Relay Output WW-54500-08 Mfr # DT-501XD-CPT
COMPARE
6-digit LED, red, 0.87\" (22 mm) H Panel Meter Tachometer, 9-35 VDC, Relay Output
₹88,100.00INR / Each
Showing 1- 7 of 7