Shimpo DT-3015N Portable Stroboscopes
Ideal for reading the object frequency when a still image appears, this stroboscope is a non-contact device that creates visuals of rotating or moving objects to observe their appearance in slow-motion
  • Internal, fast rechargeable battery supply
  • Aluminum construction provides exceptional durability
  • High polish and focused reflector eliminates "blind" viewing areas
  • External trigger allows unit to be automatically synchronized with equipment
  • Light emission auto-off feature from 1 to 120 minutes enables extended battery operation
  • Phase shift, set in degrees or ms, allows visual analysis of rotating/reciprocating objects
  • Save up to five user configurations to enable switching quickly between machinery
  • Synchronous output enables strobe to drive other strobes
  • Easy tripod or permanent mounting with integral mounting thread
This heavy duty, battery powered stroboscope possesses the added convenience of total portability via its own internal power supply. The quick recharge battery lasts up to three hours of full continuous usage with its high intensity flash rate. It provides power flexibility by utilizing either its rechargeable internal batteries or AC line-power when plugged into the included AC adapter/charger unit. These features make it ideal for quality inspection and preventive maintenance of automated motion equipment.
381,000.00 - ₹456,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flash Range (fpm) Power (VAC) Description
Availability Pricing
Shimpo Portable Stroboscope with Rechargeable Battery, 40 to 35,000 fpm 2530519 Mfr # DT-3015N
COMPARE
35000 100 to 240 Portable Stroboscope with Rechargeable Battery, 40 to 35,000 fpm
₹381,000.00INR / Each
Shimpo Portable Stroboscope Kit, 40 to 35,000 fpm, with Rechargeable Battery, Spare Flashtube, and Protective Carrying Case 2530520 Mfr # DT-3015N-KIT
COMPARE
35000 100 to 240 Portable Stroboscope Kit, 40 to 35,000 fpm, with Rechargeable Battery, Spare Flashtube, and Protective Carrying Case
₹456,000.00INR / Each