Shimpo Portable Stroboscope Kit, 40 to 35,000 fpm, with Rechargeable Battery, Spare Flashtube, and Protective Carrying Case
Representative image only.
Shimpo Portable Stroboscope Kit, 40 to 35,000 fpm, with Rechargeable Battery, Spare Flashtube, and Protective Carrying Case
Shimpo | Mfr: DT-3015N-KIT| Item: 2530520
Ideal for reading the object frequency when a still image appears, this stroboscope is a non-contact device that creates visuals of rotating or moving objects to observe their appearance in slow-motion
₹456,000.00INR / Each
₹456,000.00INR / Each
Buy It Together
Current Item
Shimpo Portable Stroboscope Kit, 40 to 35,000 fpm, with Rechargeable Battery, Spare Flashtube, and Protective Carrying Case
Shimpo Portable Stroboscope Kit, 40 to 35,000 fpm, with Rechargeable Battery, Spare Flashtube, and Protective Carrying Case
2530520
₹456,000.00INR / Each
+ =
Total Price 486000.00 ADD 2 ITEMS TO CART
Product Description
Includes
Spare flashtube, one rechargeable NiMH battery, and protective carrying case.
Key Features
  • Internal, fast rechargeable battery supply
  • Aluminum construction provides exceptional durability
  • High polish and focused reflector eliminates "blind" viewing areas
  • External trigger allows unit to be automatically synchronized with equipment
  • Light emission auto-off feature from 1 to 120 minutes enables extended battery operation
  • Phase shift, set in degrees or ms, allows visual analysis of rotating/reciprocating objects
  • Save up to five user configurations to enable switching quickly between machinery
  • Synchronous output enables strobe to drive other strobes
  • Easy tripod or permanent mounting with integral mounting thread
  • 1 year warranty
More About this Item
This heavy duty, battery powered stroboscope possesses the added convenience of total portability via its own internal power supply. The quick recharge battery lasts up to three hours of full continuous usage with its high intensity flash rate. It provides power flexibility by utilizing either its rechargeable internal batteries or AC line-power when plugged into the included AC adapter/charger unit. These features make it ideal for quality inspection and preventive maintenance of automated motion equipment.
Specifications
  • Min Flash Range (fpm)40
  • Max Flash Range (fpm)35000
  • Flash Resolution (fpm)30.0 to 5000.0 : 0.1 fpm; 5001.0 to 8000.0 to 0.2 fpm; 8001.0 to 10000.0 : 0.5 fpm; 10001.0 to 35000.0 : 1.0 fpm
  • Flash Accuracy (fpm)±0.05 % of reading
  • Flash Duration30 µsec
  • Flashtube LifeTypically 1200 hours at 1500 fpm
  • InputH level: 2.5 – 12 V; L level: 0 – 0.5 V Pulse width : 50 µs or more; Input impedance : 10 kΩ or more; 27 to 35,020 fpm (1 fpm resolution) with available delay setting in degree or msec
  • Display Type6-digit, 7 segment red LED
  • Battery LifeApprox. 3 hours (when the emission count is 1500 FPM)
  • Battery Type1 Rechargeable NiMH (included)
  • Power (VAC)100 to 240
  • DescriptionPortable Stroboscope Kit, 40 to 35,000 fpm, with Rechargeable Battery, Spare Flashtube, and Protective Carrying Case