Shimpo Digital Torque Meters
Quality control and quality assurance instrument for food, beverage, cosmetics and other torque applications
  • Sample size from 10-190mm diameter
  • Measures opening and closing torque values
  • Store 1000 data points
  • USB Interface
  • Rechargeable NiMH batteries or AC adapter
The Shimpo TNP digital torque meters are precision instruments for accurately measuring torque values of opening and closing a wide range of bottles and jars. The TNP also ensure valves, rotary switches, gear heads, torque wrenches and actuators meet requred torque specifcations.
Up to 1000 data points can be stored, along with an average value, maximum value and minimum values with the push of a single button. The automatic taring function provides quick and easy testing, with high accuracy results. A Hi-Lo comparator function tests and compares large batches of test samples to set values in an efficient and precise manner to ensure overall quality of product. A USB output port allows for interfacing with the included software package, for complete data acquisition and statistical analysi
277,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m)
Availability Pricing
Shimpo TNP-10 Digital Torque Meter; 88.5 lbf-in WW-59891-05 Mfr # TNP-10
COMPARE
88.5 10
₹277,000.00INR / Each
Shimpo TNP-2 Digital Torque Meter; 17.7 lbf-in WW-59891-06 Mfr # TNP-2
COMPARE
17.7 2
₹277,000.00INR / Each
Shimpo TNP-5 Digital Torque Meter; 44.25 lbf-in WW-59891-07 Mfr # TNP-5
COMPARE
44.25 5
₹277,000.00INR / Each
Shimpo TNP-0.5 Digital Torque Meter; 4.425 lbf-in WW-59891-08 Mfr # TNP-0.5
COMPARE
4.425 0.5