SGE forte™ Capillary Columns for Gas Chromatography

SolGel-WAX
Application Areas: Recommended for highly active compounds

Operating Temperature:
0.25-1.0µm film thickness 30°C to 260/280°C
Suitable replacement for:
DB-Wax, Rtx-Wax, Stabilwax, HP20M, HP-Wax, HP-INNOWax, Supelcowax-10, AT-Wax, Nukol, CP Wax 52CB, VB-WAX, ZB-WAX.

BP20 (Wax)

Application Areas: alcohol, free acids, fatty acid methyl esters, aromatics, solvents, essential oils

Operating Temperature:
0.1-1.0µm film thickness 20°C to 260/280°C
>1.0µm film thickness 20°C to 240/260°C

Suitable Replacement for: DB-Wax, HP-20M, Supelcowax 10, CB-Wax, Stabilwax, Carbowax, HP-Innowax, Rtx-WAX, PE-WAX, RH-WAX, ZB-WAX, TRWAX

BP21 (FFAP)

Application Areas: Volatile free acids, fatty acid methyl esters, alcohols, aldehydes, acrylates, ketones

Operating Temperature:
0.25-1.0µm film thickness 35°C to 240/250°C

Suitable Replacement for: DB-FFAP, HP-FFAP, Stabilwax-DA, CPWax-58CB

BP10 (1701)

Application Areas: Environmental analyses (EPA methods 608 and 8081), pesticides/herbicides, drugs of abuse, pharmaceuticals,

Operating Temperature:
0.25-0.5µm film thickness -20°C to 280/300°C
1.0µm film thickness -20°C to 260/280°C

Suitable Replacement for: DB-1701, Rtx-1701, SPB-7, HP-1701, CP-Sil 19CB, 007-1701, PE-1701, SP-1701

BPX-VOLATILES

Application Areas: Environmental analyses, volatile organics, alcohol analysis, USP G43

perating Temperature:
1.0-3.0µm film thickness 0°C to 290/300°C

Suitable Replacement for: DB-VRX, HP-624, OPTIMA 624, ELITE-624, 007-624, RTX-VOLATILES, SPB-624, TRV1, CPSIL 13 CB, VOCOL, VB-624, CP-624

BP624

Application Areas: EPA method 624, drinking water volatiles, chlorinated hydrocarbons, solvents
Operating Temperature:
1.0-3.0µm film thickness 0°C to 230/240°C

Suitable Replacement for: DB-624, OV-624, AT-624, HP-VOC, CP-Select624CB, 007-624, Rtx-Volatiles, VOCOL, ZB-624

CYDEX-B

Application Areas: Separation of enantiomeric compounds found in natural products

perating Temperature:
30°C to 220/240°C

Suitable Replacement for: Cyclodex-B, Rt-BDEXm
53,400.00 - ₹164,000.00INR / Each

70 variations of this product are available.

See All Product Options Below
70 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
SGE 25M X 0.22MM ID; BP10 0.25UM 1597511 Mfr # 54253
COMPARE
₹79,600.00INR / Each
SGE 30M X 0.25MM ID; BP10 0.25UM 1597512 Mfr # 54256
COMPARE
₹79,600.00INR / Each
SGE 30M X 0.32MM ID; BP10 0.25UM 1597513 Mfr # 54259
COMPARE
₹84,800.00INR / Each
SGE 25M X 0.32MM ID; BP10 0.5UM 1597514 Mfr # 54268
COMPARE
₹84,800.00INR / Each
SGE 30M X 0.32MM ID; BP10 1.0UM 1597515 Mfr # 54270
COMPARE
₹84,800.00INR / Each
SGE 25M X 0.53MM ID; BP10 1.0UM 1597516 Mfr # 54280
COMPARE
₹93,700.00INR / Each
SGE 30M X 0.53MM ID; BP10 1.0UM 1597517 Mfr # 54283
COMPARE
₹93,700.00INR / Each
SGE 10M X 0.1MM ID; BP20 0.1UM 1597522 Mfr # 54405
COMPARE
₹56,100.00INR / Each
SGE 30M X 0.25MM ID; BP20 0.5U 1597523 Mfr # 54415
COMPARE
₹83,600.00INR / Each
SGE 25M X 0.22MM ID; BP20 0.25UM 1597524 Mfr # 54421
COMPARE
₹83,600.00INR / Each
SGE 50M X 0.22MM ID; BP20 0.25UM 1597525 Mfr # 54422
COMPARE
₹144,000.00INR / Each
SGE 30M X 0.22MM ID; BP20 0.25UM 1597526 Mfr # 54424
COMPARE
₹83,600.00INR / Each
SGE 60M X 0.22MM ID; BP20 0.25UM 1597527 Mfr # 54425
COMPARE
₹144,000.00INR / Each
SGE 30M X 0.25MM ID; BP20 0.25UM 1597528 Mfr # 54427
COMPARE
₹83,600.00INR / Each
SGE 60M X 0.25MM ID; BP20 0.25UM 1597529 Mfr # 54428
COMPARE
₹144,000.00INR / Each
SGE 50M X 0.32MM ID; BP20 0.25UM 1597530 Mfr # 54431
COMPARE
₹155,000.00INR / Each
SGE 30M X 0.32MM ID; BP20 0.25UM 1597531 Mfr # 54433
COMPARE
₹89,000.00INR / Each
SGE 60M X 0.32MM ID; BP20 0.25UM 1597534 Mfr # 54434
COMPARE
₹155,000.00INR / Each
SGE 25M X 0.32MM ID; BP20 0.5UM 1597535 Mfr # 54436
COMPARE
₹89,000.00INR / Each
SGE 50M X 0.32MM ID; BP20 0.5UM 1597536 Mfr # 54437
COMPARE
₹155,000.00INR / Each
Showing 1 - 20 of 70

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat