Setra Room Pressure Monitor/Alarms
Designed for critical low differential pressure applications
  • Touch-screen display
  • Password enabled
  • Local audible alarm
  • Variable alarm delay
  • Bar graph display
These room pressure monitors provide stringent pressure monitoring and alarming. The pressure monitor can be configured to monitor the positive or negative airflow in protected environments. Touch screen allows convenient fingertip access to menus that guide you through setup and operation.
Visual red and green LEDs and a local audible alarm to alert personnel to system status. The monitor also features a SPST relay output for remote indication of system status. True differential pressure is measured and displayed with 0.001 inches of water column resolution. Additional features include multiple selectable analog outputs, output and display averaging, and door status indication.
Applications - Pharmaceutical manufacturing, Semiconductor manufacturing, Cleanrooms, Research laboratories
477,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (in WC) Min Pressure (in WC) Description
Availability Pricing
Setra SRPM-R05WB-A1-E Room Pressure Monitor/Alarm, +/-0.05" WC, 24 VAC 9807400 Mfr # SRPM-R05WB-A1-E
COMPARE
0.05 -0.05 Room Pressure Monitor/Alarm, +/-0.05" WC, 24 VAC
₹477,000.00INR / Each
Setra SRPM-0R1WB-A1-E Room Pressure Monitor/Alarm, +/-0.1" WC, 24 VAC 9807402 Mfr # SRPM-0R1WB-A1-E
COMPARE
0.1 -0.1 Room Pressure Monitor/Alarm, +/-0.1" WC, 24 VAC
₹477,000.00INR / Each
Setra SRPM-R25WB-A1-E Room Pressure Monitor/Alarm, +/-0.25" WC, 24 VAC 9807404 Mfr # SRPM-R25WB-A1-E
COMPARE
0.25 -0.25 Room Pressure Monitor/Alarm, +/-0.25" WC, 24 VAC
₹477,000.00INR / Each
Setra SRPM-001WB-A1-E Room Pressure Monitor/Alarm, +/-1" WC, 24 VAC 9807408 Mfr # SRPM-001WB-A1-E
COMPARE
1 -1 Room Pressure Monitor/Alarm, +/-1" WC, 24 VAC
₹477,000.00INR / Each
Setra SRPM-0R1WB-V1-E Room Pressure Monitor/Alarm, +/-0.1" WC, 120/240 VAC 9807412 Mfr # SRPM-0R1WB-V1-E
COMPARE
0.1 -0.1 Room Pressure Monitor/Alarm, +/-0.1" WC, 120/240 VAC
₹477,000.00INR / Each
Setra SRPM-0R5WB-V1-E Room Pressure Monitor/Alarm, +/-0.5" WC, 120/240 VAC 9807416 Mfr # SRPM-0R5WB-V1-E
COMPARE
0.5 -0.5 Room Pressure Monitor/Alarm, +/-0.5" WC, 120/240 VAC
₹477,000.00INR / Each
Setra SRPM-001WB-V1-E Room Pressure Monitor/Alarm, +/-1" WC, 120/240 VAC 9807418 Mfr # SRPM-001WB-V1-E
COMPARE
1 -1 Room Pressure Monitor/Alarm, +/-1" WC, 120/240 VAC
₹477,000.00INR / Each
Showing 1- 7 of 7