Setra Digital Barometric and Absolute Pressure Monitor
Pressure transfer standard
  • Altimeter calibration
  • Pressure switch setting
  • Min/Max tracking
  • Hi/Lo audible alarms
These barometric pressure monitors feature pressure and altitude data displayed on a 6-digit LCD; data is also accessible through a bidirectional RS-232 port. Numeric key pad provides easy access to engineering unit conversions, min/max tracking, entry of Hi/Lo alarm setpoints, nonlinear functions and calibration procedures.
Features:
0.02% full-scale accuracy
High resolution 6-digit LCD for pressure or altitude monitoring
Bidirectional RS-232 digital communications I/O port
Conversions for pressure and altitude
Digital altimeter setting indicator (DASI) and corrected altimeter mode
Programmable nonlinear functions
1,271,000.00 - ₹1,356,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Process Connection Description
Availability Pricing
Setra 370 600-1100 HPA Barometric pressure monitor, 600 to 1100 hPa/mb 9807420 Mfr # 370_6-11MB
COMPARE
±0.02% full-scale at 70°F (21°C) 1/8" NPT(F) Barometric pressure monitor, 600 to 1100 hPa/mb
₹1,356,000.00INR / Each
Setra 370 0 TO 50 PSIA Digital Absolute Pressure Monitor, 0 to 50 psia 9807428 Mfr # 370_50A
COMPARE
±0.02% full-scale at 70°F (21°C) 1/8" NPT(F) Digital Absolute Pressure Monitor, 0 to 50 psia
₹1,271,000.00INR / Each