Setra 370 0 TO 50 PSIA Digital Absolute Pressure Monitor, 0 to 50 psia
Representative image only.
Setra 370 0 TO 50 PSIA Digital Absolute Pressure Monitor, 0 to 50 psia
Setra | Mfr: 370_50A| Item: 9807428
Pressure transfer standard
₹1,271,000.00INR / Each
for an additional
₹37,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹1,271,000.00INR / Each
Buy It Together
Current Item
Setra 370 0 TO 50 PSIA Digital Absolute Pressure Monitor, 0 to 50 psia
Setra 370 0 TO 50 PSIA Digital Absolute Pressure Monitor, 0 to 50 psia
9807428
₹1,271,000.00INR / Each
+ =
Total Price 1308400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Altimeter calibration
  • Pressure switch setting
  • Min/Max tracking
  • Hi/Lo audible alarms
  • 1 year warranty
More About this Item
These barometric pressure monitors feature pressure and altitude data displayed on a 6-digit LCD; data is also accessible through a bidirectional RS-232 port. Numeric key pad provides easy access to engineering unit conversions, min/max tracking, entry of Hi/Lo alarm setpoints, nonlinear functions and calibration procedures.
Features:
0.02% full-scale accuracy
High resolution 6-digit LCD for pressure or altitude monitoring
Bidirectional RS-232 digital communications I/O port
Conversions for pressure and altitude
Digital altimeter setting indicator (DASI) and corrected altimeter mode
Programmable nonlinear functions
1 Year Warranty
Specifications
  • Accuracy±0.02% full-scale at 70°F (21°C)
  • Min Pressure (psia)0
  • Max Pressure (psia)50
  • Process Connection1/8" NPT(F)
  • Unit Of Measurementpsi, mbar, hPa, mm Hg, " Hg, mm WC, " WC
  • Display6-digit LCD with annunciators for alarms, min/max, and engineering units
  • Battery Type110/220 VAC, 50/60 Hz
  • Electrical Connectionsterminal block
  • OutputRS-232
  • Media CompatibilityClean, dry air and noncondensable gases
  • DescriptionDigital Absolute Pressure Monitor, 0 to 50 psia