Self-Contained Ice Makers
Deliver pure ice with superior technology, efficiency & reliability.
  • Built-in antimicrobial protection against mildew and fungus
  • Large bin door for easy access
  • Produces perfect ice cubes or flakes, consistent in size and shape
  • Environmentally friendly — uses non-CFC refrigerant!
These ice makers provide maximum efficiency and performance under continuous operating conditions. Designed to be energy efficient, they consume 15% less energy and 10% less water. Units turn on and off automatically to ensure the self-contained storage bin is full.
Constructed of stainless steel and featuring an air-cooled compressor, these ice makers use environmentally friendly non-CFC refrigerant and are easy to clean. Attached legs let you raise the ice maker for floor clearance and for leveling on uneven floors. Units require 15 cm of clearance on sides and back for ventilation.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Condenser Type Ice/24 hrs (lbs)
Availability Pricing
Manitowoc Ice Machine with self contained bin (55kgs in 24hrs) WW-15944-32 Mfr # EC50
COMPARE
Call For Price
Manitowoc Flake Ice Machine(83 kgs in 24 hrs.)with 27 kg self contained bin WW-15944-33 Mfr # RF0266
COMPARE
Call For Price
Manitowoc Flake Ice Machine (141 kgs in 24 hrs.) WW-15944-34 Mfr # RF0300AZ251
COMPARE
Call For Price
Manitowoc Ice Machine (204kgs in 24hrs) with Ice Bin (132 Kgs) WW-15944-35 Mfr # QD462A400
COMPARE
Call For Price
Manitowoc Flake Ice Machine (312 kgs in 24 hrs.) with Ice Bin (192 Kgs) WW-15944-36 Mfr # RF0650A570
COMPARE
Call For Price
Manitowoc Flake Ice Machine (538 kgs in 24 hrs) with Ice Bin (192 Kgs) WW-15944-37 Mfr # RF1200A570