Manitowoc Flake Ice Machine (312 kgs in 24 hrs.) with Ice Bin (192 Kgs)
Representative image only.
Manitowoc Flake Ice Machine (312 kgs in 24 hrs.) with Ice Bin (192 Kgs)
Manitowoc | Mfr: RF0650A570| Item: 1594436
Deliver pure ice with superior technology, efficiency & reliability.
Call For Price
Call For Price
Product Description
Includes
storage bin and power cord.
Key Features
  • Built-in antimicrobial protection against mildew and fungus
  • Large bin door for easy access
  • Produces perfect ice cubes or flakes, consistent in size and shape
  • Environmentally friendly — uses non-CFC refrigerant!
  • 3 year warranty
More About this Item
These ice makers provide maximum efficiency and performance under continuous operating conditions. Designed to be energy efficient, they consume 15% less energy and 10% less water. Units turn on and off automatically to ensure the self-contained storage bin is full.
Constructed of stainless steel and featuring an air-cooled compressor, these ice makers use environmentally friendly non-CFC refrigerant and are easy to clean. Attached legs let you raise the ice maker for floor clearance and for leveling on uneven floors. Units require 15 cm of clearance on sides and back for ventilation.
3 Year Warranty
Specifications
  • DescriptionFlake Ice Machine (312 kgs in 24 hrs.) with Ice Bin (192 Kgs)