Sealed electrodes for Corning Meters
Save money without giving up temperature compensation!
  • Electrodes designed to work with meters you already have
  • Find the design that works best for your application without losing automatic temperature compensation
“All-in-One” electrodes offer versatility and customization. These electrodes combine the pH reference, pH half-cell, and temperature sensor in one 12mm-diameter electrode body. The seven designs allow you to select between low-maintenance, sealed electrodes or clog-resistant refillable electrodes. Tuff-Tip electrodes feature a semi-domed tip which protects the pH bulb and ensures a quick response in harsh and dirty samples. PBR-style electrodes provide predictable, reliable, and quick responses—no matter what the sample temperature. This design virtually eliminates drift, accuracy problems, and premature performance losses due to temperature. All electrodes include 3-ft cable and BNC connector for pH, except where noted. Corning-compatible electrodes use the same temperature compensation element as Beckman, but offer an RCA plug for the temperature connection.
23,100.00 - ₹37,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
All-in-One pH/ATC Probe, Sealed/SJ/Epoxy; for Meters
₹23,100.00INR / Each
COMPARE
All-in-One pH/ATC Probe, Sealed/DJ/Epoxy; for Meters
₹30,300.00INR / Each
COMPARE
All-in-One pH/ATC Probe, Sealed/DJ/Epoxy/Tuff-Tip; Corning
₹37,400.00INR / Each