Environmental Express All-in-One pH/ATC Probe, Sealed/DJ/Epoxy; for Meters
Representative image only.
Environmental Express All-in-One pH/ATC Probe, Sealed/DJ/Epoxy; for Meters
Environmental Express | Item: 2750722
Save money without giving up temperature compensation!
₹30,300.00INR / Each
₹30,300.00INR / Each
Product Description
Key Features
  • Electrodes designed to work with meters you already have
  • Find the design that works best for your application without losing automatic temperature compensation
  • 1 year warranty
More About this Item
“All-in-One” electrodes offer versatility and customization. These electrodes combine the pH reference, pH half-cell, and temperature sensor in one 12mm-diameter electrode body. The seven designs allow you to select between low-maintenance, sealed electrodes or clog-resistant refillable electrodes. Tuff-Tip electrodes feature a semi-domed tip which protects the pH bulb and ensures a quick response in harsh and dirty samples. PBR-style electrodes provide predictable, reliable, and quick responses—no matter what the sample temperature. This design virtually eliminates drift, accuracy problems, and premature performance losses due to temperature. All electrodes include 3-ft cable and BNC connector for pH, except where noted. Corning-compatible electrodes use the same temperature compensation element as Beckman, but offer an RCA plug for the temperature connection.
1 Year Warranty
Specifications
  • ApplicationsSealed
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • Max Temperature (° C)80
  • ATC ConnectorRCA Plug
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionAll-in-One pH/ATC Probe, Sealed/DJ/Epoxy; for Meters