Scienceware Write-on Tape and Dispenser
Accepts pen, pencil, or solvent ink marker
  • Autoclavable
White autoclavable tape withstands temperatures from –99 to 250°F (–73 to 121°C). Tapes are on a 3" (7.6 cm) diameter core.
23,800.00 - ₹28,800.00INR / Pkg of 6
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware F134830050 Write-On Tape, 1/2"W x 120', Pack of 6 0827743 Mfr # F134830050
COMPARE
Write-On Tape, 1/2"W x 120', Pack of 6
₹28,800.00INR / Pkg of 6
Scienceware F134830100 Write-On Tape, 1"W x 120', Pack of 3 0827747 Mfr # F134830100
COMPARE
Write-On Tape, 1"W x 120', Pack of 3
₹23,800.00INR / Pkg of 3