Scienceware Weighted Safety Shield
Wide-angled for excellent front and side protection
Shields are constructed of 3/16" thick, clear polycarbonate to protect from splash and flying debris. Three-sided construction with epoxy-coated, steel base provides added stability and coverage.
143,000.00 - ₹152,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Height (in)
Availability Pricing
Scienceware F24962-0000 Weighted safety shield, 30" height WW-86469-10 Mfr # F24962-0000
COMPARE
30
₹143,000.00INR / Each
Scienceware F24964-0000 Weighted safety shield, 36" height WW-86469-11 Mfr # F24964-0000
COMPARE
36
₹152,000.00INR / Each