Scienceware Spinbar PTFE, Flea Micro Stir BarsSPINWEDGE STIRRING BAR, PTFE
Wedge shaped stir bar is designed to plow up sediment for thorough dispersion. Sharply angled broad face provides strong mixing action, ideal for viscious solutions.
2,100.00 - ₹3,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware F371230000 Spinwedge, Magnetic Stir Bar, 9.5 x 12.7 mm; 1/Pk WW-04775-21 Mfr # F371230000
COMPARE
Spinwedge, Magnetic Stir Bar, 9.5 x 12.7 mm; 1/Pk
₹3,200.00INR / Each
Scienceware F371230002 Spinwedge, Magnetic Stir Bar, 9.5 x 25 mm; 1/Pk WW-04775-23 Mfr # F371230002
COMPARE
Spinwedge, Magnetic Stir Bar, 9.5 x 25 mm; 1/Pk
₹2,100.00INR / Each