Scienceware F371230002 Spinwedge, Magnetic Stir Bar, 9.5 x 25 mm; 1/Pk

Representative image only.

Scienceware F371230002 Spinwedge, Magnetic Stir Bar, 9.5 x 25 mm; 1/Pk

Scienceware | Mfr: F371230002| Item: 0477523

₹3,000.00INR / Each

₹3,000.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Wedge shaped stir bar is designed to plow up sediment for thorough dispersion. Sharply angled broad face provides strong mixing action, ideal for viscious solutions.

1 Year Warranty
Specifications
  • Diameter (in)0.3125
  • Diameter (cm)0.8
  • DescriptionSpinwedge, Magnetic Stir Bar, 9.5 x 25 mm; 1/Pk
  • Warranty1 year
Reviews
Q&A