Scienceware CIRCULUS MAGNETIC STIRBARS
Offers fast mixing action and high turbulence with superior stability and less drifting at higher speeds. Ideal for highly viscous mixtures, metal particles in fluids, and containers with round or convex bottoms. End disks have solid PTFE coating and are 20mm in diameter.
3,200.00 - ₹3,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Description
Availability Pricing
Scienceware F371700000 Circulus Stirring Bar, PTFE, 32mm, White WW-04775-46 Mfr # F371700000
COMPARE
3.2 Circulus Stirring Bar, PTFE, 32mm, White
₹3,200.00INR / Each
Scienceware F371720000 Circulus Stirring Bar, PTFE, 52mm, White WW-04775-48 Mfr # F371720000
COMPARE
5.2 Circulus Stirring Bar, PTFE, 52mm, White
₹3,600.00INR / Each