Sartorius Temperature Calibration Bowls
Provides automatic calibartions
  • Designed for use with Mark 3 Series moisture analyzers
The temperature calibration bowl provides automatic calibrations for your moisture analyzer. To calibrate the internal temperature sensor, simply remove the tray support and breeze shield wihin the heating chamber, and install the bowl.
211,000.00 - ₹222,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sartorius YTM10MA Temperature Calibration Bowl for Mark-3 LTE Moisture Analyzer 1121327 Mfr # YTM10MA
COMPARE
Temperature Calibration Bowl for Mark-3 LTE Moisture Analyzer
₹211,000.00INR / Each
Sartorius YTM09MA Temperature Calibration Bowl for Mark-3 HP Moisture Analyzer 1121328 Mfr # YTM09MA
COMPARE
Temperature Calibration Bowl for Mark-3 HP Moisture Analyzer
₹222,000.00INR / Each